Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IgG2b Kappa Light Chain Isotype Control (MG2b), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP143317R
Description
IgG2b Kappa Light Chain Monoclonal specifically detects IgG2b Kappa Light Chain in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Control.Specifications
IgG2b Kappa Light Chain | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Affinity Purified | |
Isotype Control (Primary) | |
Store at 4°C in the dark. |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
MG2b | |
Flow Cytometry : 1:10 - 1:1000, Immunohistochemistry : 1:10-1:500, Immunocytochemistry/Immunofluorescence : 1:10-1:500, Control | |
Mouse | |
0.5 mL | |
Mouse | |
IgG2b κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction