Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IgG2b Negative Control Mouse, Clone: GC198, Isotype Control, MilliporeSigma™
Mouse Isotype Control
Supplier: MilliporeSigma MABC006
Description
Mouse IgG2b Negative Control Monoclonal Antibody validated for use in Flow Cytometry and Immunofluorescence.Specifications
IgG2b | |
Monoclonal | |
0.5mg/mL | |
Purified from mouse ascitic fluid. The antibody is supplied in 0.5mL 25mM Tris-HCl, 0.4M NaCl, pH 8.0, containing 0.2% bovine serum albumin and 0.1% sodium azide. The characteristics of each lot are tested by electrophoresis, specific immunofluorescence | |
Protein I preparation from Neisseria gonorrhoeae. | |
Primary | |
Purified |
Flow Cytometry, Immunofluorescence | |
GC198 | |
Unconjugated | |
Mouse | |
500 μL | |
Store at 2 to 8°C for up to 6 months. For prolonged periods store below -20°C in undiluted aliquots. AVOID REPEATED FREEZE/THAW CYCLES. WARNING: The monoclonal reagent solution contains 0.1% sodium azide as a preservative. |
|
IgG2b |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction