Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IgG1 Isotype Control, PE
Mouse Isotype Control
Supplier: Invitrogen MG104
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
This item is not returnable.
View return policy
Description
Mouse IgG1 Isotype Control for Flow
Mouse IgG1 is a subtype of immunoglobulin G antibody that is produced by B cells in response to an antigen. It is the most abundant IgG subtype in the serum and plays a crucial role in the adaptive immune response. Mouse IgG1 is composed of two heavy chains and two light chains, and it is involved in opsonization, complement activation, and antibody-dependent cell-mediated cytotoxicity. It is also involved in the regulation of immune responses, including the activation of T cells and the inhibition of B cell activation. Mouse IgG1 is widely used in research and diagnostic applications, including ELISA, Western blotting, and immunohistochemistry. It is also used in therapeutic applications, such as immunotherapy and vaccine development.![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Mouse IgG1 | |
0.1mg/mL | |
PBS with 0.1% sodium azide | |
Mouse | |
500 μL | |
Whole Antibody | |
1 Each | |
Not Applicable | |
Liquid | |
50 Tests |
Flow Cytometry | |
PE | |
IgG1; Igh-4; Ighg1; Immunoglobulin G; Immunoglobulin G1; immunoglobulin heavy constant gamma 1 (G1m marker); ImmunoglobulinG; ImmunoglobulinG1; VH7183 | |
purified | |
ASR | |
Primary | |
0.1% Sodium azide | |
2-8°C | |
IgG1 | |
Wet Ice |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Analyte Specific Reagent. Analytical and performance characteristics are not established.
Spot an opportunity for improvement?
Provide Content Correction