Learn More
Mouse IgG1 Negative Control Antibody, Mouse, Clone: 1E2.2, Isotype Control, MilliporeSigma™
Mouse Isotype Control
Supplier: MilliporeSigma CBL610
Description
Mouse IgG1 Negative Control Monoclonal Antibody validated for use in Flow Cytometry.Specifications
IgG1 | |
Monoclonal | |
1mg/mL | |
Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
Protein G | |
Primary | |
Purified |
Flow Cytometry | |
1E2.2 | |
Unconjugated | |
Mouse | |
100 μg | |
2-8°C for one year | |
IgG1 κ |