Learn More
PBL Assay Science VeriKine-HS Mouse Interferon Beta (IFN-Beta, IFNb) ELISA, Plasma/Serum/Tissue Culture Media (TCM) (High Sensitivity) [5x96 well plate]
Supplier: PBL Assay Science 424102
VeriKine-HS Mouse Interferon Beta ELISA is manufactured by PBL Assay Science (Catalog #42410-2) for research use only (RUO) and packaged as a 5x96-well plate. This assay is serum, plasma and tissue culture media (TCM) compatible and has an assay range of 0.94-60 pg/mL. This ELISA kit has intra- and inter-assay CVs of less than 10%.