Learn More
PBL Assay Science VeriKine Mouse Interferon Alpha (IFN-Alpha, IFNa) ELISA, Tissue Culture Media (TCM) [1x96 well plate]
Supplier: PBL Assay Science 421201
VeriKine Mouse Interferon Alpha ELISA is manufactured by PBL Assay Science (Catalog #42120-1) for research use only (RUO) and packaged as a 1x96-well plate. This assay detects all 14 Mouse IFN-Alpha subtypes in TCM and has an assay range of 12.5-400 pg/mL. This ELISA kit has intra- and inter-assay CVs of less than 10%.