Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Morphine, Mouse, Clone: BDI263, Abnova™
Mouse monoclonal antibody raised against Morphine.
Supplier: Abnova Corporation MAB4261
Specifications
Morphine | |
Monoclonal | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
Mouse | |
1 mg | |
Primary | |
Store at 4°C. | |
IgG1 λ |
ELISA | |
BDI263 | |
Mouse monoclonal antibody raised against Morphine. | |
In 150mM PBS, pH 7.4 (0.09% sodium azide) | |
Morphine conjugated with BTG. | |
RUO | |
Reacts with Morphine and Codeine. Cross reactivity was determined by ELISA with competition assays. Does not react with Nalorphine, Meperidine or Methadone. | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction