Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Influenza A virus nucleoprotein, Mouse, Clone: InA224, Abnova™
Mouse monoclonal antibody raised against influenza A virus nucleoprotein.
Supplier: Abnova Corporation MAB3149
Specifications
Influenza A virus nucleoprotein | |
Monoclonal | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
Mouse | |
200 μg | |
Primary | |
Virus | |
Liquid |
ELISA | |
InA224 | |
Mouse monoclonal antibody raised against influenza A virus nucleoprotein. | |
In PBS, pH 7.4 (0.09% sodium azide) | |
Influenza virus type A (strain H1N1). | |
RUO | |
This antibody detects nucleoprotein of Influenza A with high specificity and have no cross reactsivity to nucleoprotein of Influenza B virus. | |
Store at 4°C. | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction