Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse anti-IgG2a, κ, Isotype Control, APC-R700, Clone: G155-178, BD
Supplier: BD Biosciences 565126
Description
Specifications
IgG2a, κ | |
Monoclonal | |
APC-R700 | |
Mouse | |
Affinity Purified | |
RUO | |
IgG2a κ |
Flow Cytometry | |
G155-178 | |
Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide. | |
TNP-keyhole limpet hemocyanin | |
0.1 mg | |
Store undiluted at 4°C. Do not freeze. Protect from light. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction