Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dengue virus Types 1,2,3,4, Mouse, Clone: BDI419, Abnova™
Mouse monoclonal antibody raised against Dengue virus Types 1,2,3,4.
Supplier: Abnova Corporation MAB4043
Specifications
Dengue virus Types 1,2,3,4 | |
Monoclonal | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
Mouse | |
1 mg | |
Primary | |
Virus | |
Liquid |
Dot Blot, ELISA, Immunofluorescence | |
BDI419 | |
Mouse monoclonal antibody raised against Dengue virus Types 1,2,3,4. | |
In PBS, pH 7.2 (0.09% sodium azide) | |
2, 3 and 4., Mixture of Dengue 1 | |
RUO | |
Reacts with each dengue serotype in dot blot, ELISA and fluorescent antibody tests. The reactivity of this antibody varies between strains and various isolates, therefore, a single dilution may not be appropriate for each situation. The myeloma cell line used as a fusion partner produces small amounts of IgG1, therefore IgG1 is sometimes present in the IgG2a, k preparation. | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG2a κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction