Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Biotin, Mouse, Clone: SB58c, Abnova™
Mouse monoclonal antibody raised against BIOT.
Supplier: Abnova Corporation MAB3274
Specifications
Biotin | |
Monoclonal | |
Unconjugated | |
Immunofluorescence (1 ug/106 cells) ELISA (1 ug/mL) The optimal working dilution should be determined by the end user. | |
Mouse | |
500 μg | |
Primary | |
Store at 4°C. | |
IgG1 |
ELISA, Flow Cytometry | |
SB58c | |
Mouse monoclonal antibody raised against BIOT. | |
In 100mM BBS, pH 8.2 | |
KLH1139-biotin. | |
RUO | |
Biotin. | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction