Learn More
Kamiya Biomedical Mouse Microalbumin ELISA. 96 Tests.
Supplier: Kamiya Biomedical KT343
Mouse Microalbumin ELISA. 96 Tests. For the quantitative measurement of microalbumin in mouse biological samples. Assay Range: 7.81 - 500 ng/mL. Includes calibrators and control.