Learn More
Cole-Parmer Davis Instruments 7716 Mounting Tripod for Weather Stations, 5.8 ft tall
Supplier: Cole-Parmer 8640370
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Use a mounting tripod for your Davis Vantage Pro2 or Vantage Vue to make installation even easier. Includes two 3-foot (0.92-meter) poles, which may be used separately or together to make a single 5.8-foot (1.77-meter) pole.