Learn More
Mott Corporation MOTT SPARGER 20UM TIPS
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Mott Corporation 124245901200OF
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
1240S-.317-.093-1.037-20-A-OFCLS100S=ORING GROOVE, M5x.8 THDOD=0.317"ID=0.093"POROUS LENGTH = 0.800"MEDIA GRADE 20 OAL= 1.037" LONG 316L SS