Learn More
Mott Manufacturing Sink Support Kit

The Mott Sink Support Kit includes two durable metal frame supports for use with any Mott sink units.
$121.30
Catalog Number | Mfr. No. | Location | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Location | Price | Quantity & Availability | |||||
04-000-002
![]() |
Mott Manufacturing
SSK1036NE |
Zone 4 |
Each for $121.30
|
|
|||||
04-000-089
![]() |
Mott Manufacturing
SSK1036MGC |
Zone 7 |
Each for $121.30
|
|
|||||
04-000-524
|
Mott Manufacturing
SSK1036SB |
Zone 10 |
Each for $121.30
|
|
|||||
04-000-785
|
Mott Manufacturing
SSK1036CC |
Zone 1 |
Each for $121.30
|
|
|||||
04-000-176
|
Mott Manufacturing
SSK1036DT |
Zone 5 |
Each for $121.30
|
|
|||||
Description
- Two metal support frames
- Provide extra reinforcement to hold sink securely
- Zone 1 (Use CC suffix): Montana (Eastern), Minnesota, North Dakota, South Dakota, Wyoming
- Zone 2 (Use WJ suffix): Iowa, Kansas, Missouri, Nebraska, Wisconsin
- Zone 3 (Use SL suffix): Alabama, Florida, Georgia, Illinois, Kentucky, Tennessee
- Zone 4 (Use NE suffix): Connecticut, Delaware, District of Columbia, Maine, Maryland, Massachusetts, New Hampshire, North Carolina, Rhode Island, South Carolina, Vermont, Virginia, West Virginia
- Zone 5 (Previous DT suffix): Moved to Zone 9
- Zone 6 (Previous CF suffix): Moved to Zone 9
- Zone 7 (Use MGC suffix): Arizona, Arkansas, Colorado, Louisiana, Mississippi, New Mexico, Oklahoma, Texas
- Zone 8 (Use S suffix): New Jersey, New York, Pennsylvania (Eastern)
- Zone 9 (Use SV suffix): Michigan, New York State (Upper), Ohio, Pennsylvania (Western)
- Zone 10 (Use SB suffix): Alaska, California, Hawaii, Idaho, Montana (Western), Nevada, Oregon, Utah, Washington
Order Info
When ordering, be sure to select items with the letter code suffix that matches the state that items are to be DELIVERED to
Specifications
Any Mott sink unit |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.