Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Motic™ Moticam 4000 Camera
The Moticam 4000 camera is designed for ease of use and the ability to output fast live images at 4K (3840 x 2160) pixels in full HD.
Supplier: Motic™ 1100600101521
This item is not returnable.
View return policy
Description
- C-mount for lens
- MotiConnect software is available for free download (with free updates included)
- Capture Resolution: 8MP (3840 x 2160 px)
Specifications
Microscope Camera | |
8 MP | |
1/1.8 in. |
12 V Power Adapter, CS Ring, Motic™ 4-dot Calibration Slide, HDMI Cable, Manual, 12 mm Lens, 30 mm to 38 mm Adapter, Macro Tube | |
IMX334 | |
20 fps |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction