Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
United Scientific™ Mortar & Pestle Set, Porcelain, Fully Glazed
![SDP](/content/dam/fishersci/glyphs/sdp.png)
![SDP](/content/dam/fishersci/glyphs/sdp.png)
![SCI_ED](/content/dam/fishersci/glyphs/scied.png)
United Scientific™ Mortar & Pestle Set feature deep form mortars with lips are fully glazed on the inside and outside.
Supplier: United Scientific™ PMG125
This item is not returnable.
View return policy
Description
Deep form mortars with lips are fully glazed on the inside and outside. Oversized pestles are also fully glazed to grinding surface.
- Tolerance is ±5 mm
- Offered in 80 mL, 125 mL, 300 mL, and 500 mL Capacity
Specifications
Glazed | |
125 mL | |
Mortar and Pestle | |
Porcelain |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction