Learn More
Molecular Dimensions MORPHEUS HT-96
Supplier: Molecular Dimensions MD147
Protein Crystallography. Morpheus screen HT-96 (96 x 1.1mL): Simple and effective 3D grid design covering a range of pH, PEGs and salt additives.49 low molecular weight ligands promote both initial crystal formation and lattice stability.Ideal for membrane proteins. All conditions are cryo-protected.