Learn More
Fine Science Tools Moria Bulldog Clamps, Jaw: 16 x 2.5mm, 53 mm
Supplier: Fine Science Tools 1837443
Product: Moria Bulldog Clamp - Straight/53mm/ Shape: Straight/ Jaw Dimensions: 16 x 2.5mm/ Clamping Pressure: Strong/ Alloy: Stainless Steel/ Length: 53mm/ Serrated: No/ Unit: Each