Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MOPS, Biological Buffer, 99%, Spectrum™ Chemical
SDP

MO102, 1132-61-2, C7H15NO4S

Supplier:  Spectrum Chemical Manufacturing Corporation MO1021KG

 View more versions of this product

Catalog No. 18-604-696


Add to Cart

Description

Description

Spectrum™ Chemical MOPS, Biological Buffer
Specifications

Specifications

1132-61-2
0.01
1 kg
Reagent
1
Poly Bottle
99%
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit