Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Monomethyl fumarate, Thermo Scientific™
$51.65 - $187.00
Specifications
Chemical Name or Material | Monomethyl fumarate |
---|---|
CAS | 2756-87-8 |
Melting Point | 145°C to 150°C |
Shipping Condition | No special requirements |
Product Line | Building blocks |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AC469750050
|
Thermo Scientific Chemicals
469750050 |
5 g |
Each
for $51.65
Save 14% Reg :
Order in Bulk
Request bulk or custom formats |
|
|||||
AC469750250
|
Thermo Scientific Chemicals
469750250 |
25 g |
Each for $187.00
Order in Bulk
Request bulk or custom formats |
|
|||||
Description
Specifications
Monomethyl fumarate | |
145°C to 150°C | |
Building blocks | |
Powder or Crystals | |
≥96.0% |
2756-87-8 | |
No special requirements | |
C5 H6 O4 | |
White to Almost-white |
Safety and Handling
Exclamation mark
Corrosion
EINECSNumber : 220-412-6
Recommended Storage : Normal conditions
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title