Learn More
United Scientific™ Monocotyledon Stem Model
This model is patterned on a corn stem to show the general anatomy of a monocot in both longitudinal and cross sections.
Supplier: United Scientific™ MBMS01
This item is not returnable.
View return policy
Description
- Features such as the cuticle layer, epidermis, closed vascular bundles, parenchyma cells, and others are clearly shown
- This injection-molded model is more durable than the more common fiberglass construction
- Detailed key included
Specifications
Monocot Model | |
Plastic |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Warning: Cancer and reproductive harm
Prop 65