Learn More
Electron Microscopy Sciences AirChek Air Monitoring Badge, Formaldehyde
Supplier: Electron Microscopy Sciences 6447210
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Airchek Badges provide an innovative, simple and effective system for monitoring vapor levels, including formaldehyde, glutaraldehyde, xylene, toluene, and isopropanol. The badges are simply clipped on to the technician's clothing and a record is kept of name, location and time, providing continuous sampling for personal monitoring and allowing the measurements required under Government regulations.Overall Length is 8 3/4" (213 mm). Overall Length is 8 3/4" (213 mm).
One badge provides continuous sampling over 8 hours. A second badge can be used to supply information for analysis of 15-minute peak exposure periods.
Use: Formaldehyde
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.