Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Molecular sieves, 4A
$81.90
Specifications
Quantity | 250 g |
---|---|
CAS | 70955-01-0 |
Physical Form | Solid |
MDL Number | MFCD00131613 |
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
AA8795630
|
Thermo Scientific™
8795630 |
250 g | 250 g |
Each for $81.90
Order in Bulk
Request bulk or custom formats |
|
AA8812030
|
Thermo Scientific™
8812030 |
250 g | 250 g | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Specifications
250 g | |
70955-01-0 | |
Solid | |
MFCD00131613 |
Safety and Handling
ShelfLife : 5 Years
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title