Learn More
MFG Tray Nesting Containers
Nest when empty, stack on lids when full
Supplier:  MFG Tray 9231085280
 View more versions of this product
Catalog No. 15-239-58
Add to Cart
Description
Description
  • Impervious to radiation, cutting oils, greases, solvents, detergents, mild acids and alkaline solutions within a pH rating of 3.0 to 10.0
  • Temperature rating of -51° to +121°C (-60° to +250°F)
  • Smooth outside surface and radial edges
  • Tough composite construction
  • Nesting design saves space
Machine integration and robotic applications
Specifications
Specifications
Freezer Container
Red
Smooth finish
Radial Edges
9.75 x 9.25 x 4.5 in. (24.76 x 23.5 x 11.43 cm)
9 in.
22.86 cm
21.33 cm
4.4 in.
11.17 cm
-51°C to +121°C
Fiberglass
Rectangular
Machine Integration and Robotic Applications
1 Ea.
9.75 in.
24.76 cm
8.4 in.
4.5 in.
11.43 cm
-60°F to +250°F
Product Suggestions
Product Suggestions
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Lid is sold separately.