Learn More
Hach Company Ohaus Moisture Analyzer Balance, 120g Capacity
Supplier: Hach Company 2946402
The Ohaus Moisture Analyzer incorporates state-of-the-art halogen heating with precision weighing technology to quickly, effectively and affordably measure moisture content. Powerful software facilitates ease of use, as well as data storage and management. 120g model displays % moisture, % solids, Time, Weight, Temperature, Test ID and Drying Curve. This item features a 2-year manufacturer warranty. Ideal applications include wastewater treatment, food and beverage, chemical, industrial, farming, construction and research labs. Pan size: 3.5 in (90 mm)
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.