Learn More
Scientific Industries MOISTURE ANALYZER, 120G
Supplier: Scientific Industries AGS120
Moisture Analyzer - Professional, 0.01% [120g x 0.001g] 250C with 250W of drying power up to 250C, heavy duty all metal housing design, precision AG electromagnetic load cell, equipped with USB, RS232, and PS2 ports which allow for quick and stable PC connection.