Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LifeSecure™ MobileAid™ SchoolGuard™ 50-Student Trauma First Aid Kit



Comprehensive 255-piece Trauma First Aid Kit for school groups of up to 50 students.
Supplier: LifeSecure™ 37100
Description
- For a rapid response to any medical emergency
- Wide-mouth, slim-design medical duffel
- 8 pockets and pouches and interior main compartment organizer insert
- Organized with extensive first aid supplies for rapid response effective treatment of traumatic injuries in an emergency or disaster on or off campus
Specifications
Trauma First Aid Kit | |
55.88 cm | |
Red | |
Up to 50 People | |
27.94 cm |
Trauma First Aid | |
ISEA Z308.1-2015 Class A, Type II | |
1 Ea. | |
22 x 11 x 11.5 in. | |
29.21 cm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title