Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diversified Spaces™ Mobile Storage Cabinet
SCI_ED

Rolling unit aids in easy transport of supplies.

Supplier:  Diversified Spaces™ WMSC3135

Catalog No. S42454


Add to Cart

Description

Description

  • Constructed from solid maple and maple veneer
  • Requires minimal assembly
  • 4.45cm (1.75 in.) thick maple top
  • Features 0.635cm (0.25 in.) pegboard and nine hooks for tool mounting on right side of cabinet
  • Heavy duty 10.16cm (4 in.) locking swivel casters
  • Wood finished with durable chemical resistant non-emitting UV finish
  • SEFA compliant and MAS Certified Green
Specifications

Specifications

60.96 x 91.4 x 92.71 cm (24 x 36 x 36.5 in.)
Maple
Yes
Locking double doors with adjustable shelf
Limited Lifetime
Mobile Storage Cabinet
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit