Learn More
Enzo Life Sciences Matrix Metalloproteinase-2 (MMP-2) fluorometric drug discovery kit (1x96wells)
Supplier: Enzo Life Sciences BMLAK4090001
A QUANTIZYME Assay System. The MMP-2 Fluorometric (also known as fluorimetric) Drug Discovery Kit is a complete assay system designed to screen inhibitors of matrix metalloproteinase-2 (MMP-2, gelatinase A) using a quenched fluorogenic peptide: OMNIMMP fluorogenic substrate Mca-Pro-Leu-Gly-Leu-Dpa-Ala-Arg-NH. Mca fluorescence is quenched by the Dpa group until cleavage by MMPs at the Gly-Leu bond separates the two moieties. The assays are performed in a convenient 96-well microplate format. The kit is useful to screen inhibitors of MMP-2, a potential therapeutic target. Included are active enzyme, assay buffer, a prototypic control inhibitor (NNGH3), and a detailed instruction booklet. Alternative Name: Gelatinase A, 72 kDa Type IV collagenase. Applications: Fluorescent detection, HTS, Activity assay. Contents: MMP-2 enzyme (human) (recombinant), substrate, calibration standard, inhibitor, assay buffer, 96-well white microplate.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.