Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ ML 277
Selective Kv7.1 (KCNQ1) potassium channel activator; augments IKs current
$338.50 - $1404.00
Specifications
Purity | >98% |
---|---|
Product Type | ML 277 |
Molecular Weight (g/mol) | 471.59 |
Inhibitors | ML 277 |
Description
Selective Kv7.1 (KCNQ1) potassium channel activator (EC50 = 260 nM). Exhibits >100-fold selectivity versus KCNQ2, KCNQ4 and hERG potassium channels. Augments IKs current of cultured human cardiomyocytes and shortens action potential duration.Specifications
>98% | |
ML 277 | |
471.59 | |
ML 277 |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title