Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ML 171, Tocris Bioscience™
Click to view available options
Quantity:
50mg
Description
NADPH oxidase 1 (NOX1) inhibitor; blocks NOX1-dependent ROS generation (IC50 = 0.25 μM in a HEK293-NOX1 recombinant cell system). Selective for NOX1 over other NADPH oxidases (IC50 values are > 3 μM). Inhibits SrcYF-induced invadopodia formation in human DLD1 colon cancer cells.
Specifications
Quantity | 50mg |
Formula Weight | 241.31 |
Percent Purity | >98% |
Chemical Name or Material | ML 171 |
Safety and Handling
Recommended Storage : Store at +4°C