Learn More
Holland Safety Equipment MK2 FUME HOOD ALARMS
Supplier: Holland Safety Equipment M52TEL53US
The AFA500 is an airflow monitor that provides an audible and visual alarm if face velocity goes below the alarm set point. Included are all parts for a surface mount installation, incl. mounting box. Easy to field install and calibrate.