Learn More
Medchemexpress LLC HY-101869 5mg Medchemexpress, MK-0343 CAS:233275-76-8 Purity:>98%
Supplier: Medchemexpress LLC HY1018695MG
Medchemexpress, HY-101869 5mg MK-0343 CAS:233275-76-8 MK0343 (MRK-409) is an orally bioavailable GABAA receptor subtype-selective partial agonist. MK0343 is a non-sedating anxiolytic. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.