Learn More
Sklar Instruments Mixing/Solution Bowl, Sklar, Stainless Steel. This bowl measures 8-9-3/4 in. x 4 in. and has a capactiy of 3 quarts. EA/1.
SDP
Supplier:  Sklar Instruments 101492
Encompass_Preferred
Mixing/Solution Bowl, Sklar, Stainless Steel. Sklar Mixing/Solution Bowls are ideal for mixing and containing solutions required in surgical procedures. They may be found in emergency departments where wound cleaning is necessary, or operating rooms where irrigation is required for specific procedures. The top and lip of the mixing bowl does not angle outward such as that of a basin may. This bowl measures 8-9-3/4 in. x 4 in. and has a capactiy of 3 quarts.
Catalog No. 50-143-7001