Learn More
Sklar Instruments Mixing/Solution Bowl, Sklar, Stainless Steel. This bowl measures 8-1/8 in. x 3 in. and has a capactiy of 1-5/8 quarts. EA/1.
Supplier: Sklar Instruments 101491
Mixing/Solution Bowl, Sklar, Stainless Steel. Sklar Mixing/Solution Bowls are ideal for mixing and containing solutions required in surgical procedures. They may be found in emergency departments where wound cleaning is necessary, or operating rooms where irrigation is required for specific procedures. The top and lip of the mixing bowl does not angle outward such as that of a basin may. This bowl measures 8-1/8 in. x 3 in. and has a capactiy of 1-5/8 quarts.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.