Learn More
Cole-Parmer Mixer Motor; 120/230 VAC
Supplier: Cole-Parmer 5500808
Replacement parts keep your overhead mixer running smoothlyRequires SP (55008-06) or SD (55008-07) Mixer ControllerMotor capabilities are based on the controller. The mixer motor has through-shaft capability and features a user replaceable keyless chuck. Bracket hardware is preinstalled to enable installation of the adjustable tilt mounting bracket.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.