Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aqua Solutions Mixed Alcohol Solvent Prepared to ASTM D 3230-13 (4L)
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Aqua Solutions 59124L
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Mixed Alcohol Solvent Prepared to ASTM D 3230-13 (4L)
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction