Learn More
Enzo Life Sciences MITO-ID Green detection kit (100 tests)
SDP
Supplier:  Enzo Life Sciences ENZ510220100
Encompass_Preferred
Photostable,  non-toxic and selective mitochondrial dye that stains regardless of membrane potential. MITO-ID Green Detection Kit contains a proprietary membrane-permeable mitochondria-selective dye suitable for use with live-, detergent-permeabilized- and aldehyde-fixed-cells. Unlike conventional dyes (such as DiOC6(3), JC-1, rhodamine 123 and tetramethylrhodamine ethyl ester) MITO-ID Green dye highlights mitochondria regardless of their energetic state. The dye is compatible with most fluorescence detection systems, including conventional and confocal fluorescence microscopes as well as High Content Screening (HCS) platforms. The kit is useful for assessing mitochondrial morphology changes, estimating mitochondrial mass and co-localizing RFP-tagged proteins to the mitochondrial compartment. Applications: Fluorescence microscopy, Fluorescent detection.
Catalog No. 50-210-0557