Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mitchell Plastics Re-usable, Heavy Duty Waste Containers, Floor Models
Specifications
Closure Type | Hinged Lid |
---|---|
Color | Clear |
For Use With (Application) | Ideal for keeping sharp objects and liquid waste contained in the event of waste box leakage, disposing of biohazardous, as well as some radioactive materials |
Material | Acrylic |
Product Type | Floor Model Waste Container |
Catalog Number | Mfr. No. | Height (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Height (Metric) | Price | Quantity & Availability | |||||
05-723-432
|
Mitchell Plastics
WC200 |
52.70 cm | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
05-723-431
|
Mitchell Plastics
WC150 |
63.5 cm | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
05-723-434
|
Mitchell Plastics
WC150C |
63.5 cm | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Specifications
Hinged Lid | |
Ideal for keeping sharp objects and liquid waste contained in the event of waste box leakage, disposing of biohazardous, as well as some radioactive materials | |
Floor Model Waste Container |
Clear | |
Acrylic |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title