Learn More
Perkin Elmer US LLC MIRacle ATR Heated Crystal Plate - Diamond/ZnSe, 60°C
Supplier: Perkin Elmer US LLC L1272314
The MIRacle ATR accessory analyzes solids, liquids, pastes, gels, and intractable materials. In its most popular configuration, it acts as a single reflection ATR accessory with high IR throughput, which makes it ideal for sample identification and QA/QC applications. The easily changeable crystal plate design provides an analysis of a broad spectrum of sample types, while ensuring a constant sampling path length.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.