Learn More
Cole-Parmer MiniVac Waste Aspiration System, 1 L
Supplier: Cole-Parmer 0439619
These compact waste aspiration systems replace the need for glass waste containers. Durable components are easily cleaned with 70% isopropyl alcohol. Station may be used as a secondary overflow to an existing system, or as compact stand-alone unit—simply connect in line with the primary container, or to a house or pump supplied vacuum line.