Learn More
Chemglass Life Sciences Mini Run Gel Electrophoresis System
Supplier: Chemglass Life Sciences CLS190501
The Mini-Run horizontal gel apparatus can be used for almost any agarose gel electrophoresis application. The chamber has a built-in DC power supply so you can just plug it in and be ready to run your gel. The cover and gel chamber are both made from an advanced etch-resistant plastic and the electrodes are platinum coated for clean operation and durability. Power: AC100V-110V, 50-60Hz. Output Voltage: DC50V/DC100V. Bath Volume: 250mL (Each Chamber). Comb Sizes (Width x Thickness x No. of Teeth): 4mm x 1mm x 8, 4mm x 1mm x 17, 6mm x 1mm x 6, 6mm x 1mm x 12. Gel Migration Area: 60 x 55 mm or 110 x 55mm.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.