Learn More
Electron Microscopy Sciences miniLIGHT Set 3 2W
SDP
Supplier:  Electron Microscopy Sciences 7166603
Encompass_Preferred
Constructed from the highest quality material and small as its name, this miniLight offers the most universal light source, handling many tasks that require a bright light but with a small foot print that fits into most locations.
The miniLight's main component is the miniLamp. This is a small but powerful light source. It shines very bright on a small area. It has a low dissipation loss with no hot part. Long life because the lifetime of a LED is 50,000 hours. Little power needed (1watt). Adjustable intensity and CE compatible.
Catalog No. 50-346-37