Learn More
World Precision Instrument MiniFlow - Flow-through Cell with integrated planar oxygen sensor
SDP
Supplier:  World Precision Instrument 501642
Encompass_Preferred
The MiniFlow oxygen probe is a miniaturized fiber optic chemical sensor integrated in a T-shape flow through cell. The standard T-shape flow cell can be easily connected via Luer-Lock adapters to external tubings. Liquids (e.g., water, blood, etc.) can be pumped through the cell. The sensor has a response time (t90) of approximately 40-s and an excellent long-term stability.
Catalog No. 50-252-1410
Explore available promotions