Learn More
Greiner Bio-One MiniCollect™ PIXIE Heel Incision Safety Lancet
Safety lancet designed for capillary blood collection by heelstick
Supplier:  Greiner Bio-One 450525
 View more versions of this product
Catalog No. 22-405-177
Add to Cart
This item is not returnable. View return policy
Description
Description
  • Ergonomic design with non-slip, textured surface
  • Non-detachable safety tab to ensure no waste is left behind
  • One-handed activation for ease of use
  • Trigger orientation to minimize risk of tissue compression
TRUSTED_SUSTAINABILITY
Specifications
Specifications
0.65 mm
50 Pk., 200/Cs.
Micro-preemie
Yes
1.4 mm
Green
Heelstick Lancet
Safety and Handling
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov