Learn More
Research Products International Corp Mini-Blotting Containers, 15.4 x 10.4 x 2.5cm, Clear, 4 per Case
SDP
Supplier:  Research Products International Corp 248718
Encompass_Preferred
Plastic containers for many laboratory applications. Containers have a smooth, flat surface that is ideal for blotting membranes or staining gels. Hinged lid with snap lock closure prevents evaporation of reagents. Constructed of ultra-clear polystyrene.
Catalog No. 50-550-303