Learn More
Cole-Parmer Digi-Sense Miniature Type-K Thermocouple Male Connector, 2 Pin
Supplier: Cole-Parmer 1852689
Miniature connectors are suited for small wires and mineral insulated cables. Glass-filled nylon construction with hollow pins and molded barriers prevent short circuit. Secure thermocouple wire into positive/negative terminals with screws. Each connector comes in ANSI color coded material: type J-black, type K-yellow, and type T-blue.