Learn More
Key Scientific Products Mini Slide Warmer, 10.25" x 8" x 3" H 220V
Supplier: Key Scientific Products CY212972
SLIDE WARMER - MINI: Ideal for use in the fields of cytology, histology, pathology, and biology for paraffin tissue section mounting. Thermal heater ensures even heat transfer. Anodized black surface provides contrast. LED temperature display. Power Requirement: 100WSurface Size 10″ x 7″ (26 x 18 cm)Capacity: 23 slidesTemperature Range: room temperature to 70 degrees C +/- 3 degreesOverall Size: 10.25" x 8" x 3" H
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.