Learn More
Research Products International Corp Mini-Pipettor, 10ul, White
Supplier: Research Products International Corp 347020
Compact and engineered for precision dispensing, the sturdy Minipet air-displacement micropipettes come in a range of volumes (5ul to 200ul) and color options. Most important, the MiniPet micropipette provides as low as +/-3% accuracy and 2% precision at full stroke. Built from sturdy molded plastic, the air-displacement MiniPet micropipette handles thousands of pipettings.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.